accessibility menu, dialog, popup

UserName

CAS RN 3518-88-5

IUPAC Name: ethyl 3-[(3-ethoxy-3-oxopropyl)amino]propanoate
Synonyms: ethyl 3-[(3-ethoxy-3-oxopropyl)amino]propanoate
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: ONEIYJQBXMVMKU-UHFFFAOYSA-N
SMILES: CCOC(=O)CCNCCC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)