missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3

Catalog No. p-3778251
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC346930050 5 g Glass bottle
AC346930250 25 g Glass bottle
AC346931000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC346930050 Supplier Thermo Scientific Chemicals Supplier No. 346930050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15110-17-5 | C11H25O5P | 268.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 15110-17-5
Molecular Formula C11H25O5P
Molecular Weight (g/mol) 268.29
MDL Number MFCD00800259
InChI Key WKPXMGJOGYEGNW-UHFFFAOYSA-N
Synonym diethyl 3,3-diethoxypropyl phosphonate, diethyl 3,3-diethoxypropylphosphonate, phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester, 3-diethoxyphosphoryl-1,1-diethoxy-propane, 3-diethoxyphosphoryl propanaldehyde diethyl acetal, phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester
PubChem CID 328856
IUPAC Name 3-diethoxyphosphoryl-1,1-diethoxypropane
SMILES CCOC(CCP(=O)(OCC)OCC)OCC

Specifications

Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4320
Quantity 5 g
Stabilizer 1% max. Na2CO3
Formula Weight 268.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl (3, 3-diethoxypropyl)phosphonate, Stabilized with max.1% (w/w) Na2CO3
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only