missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate 96.0+%, TCI America™

Catalog No. D24385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24385G 5 g
D24381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24385G Supplier TCI America Supplier No. D24385G
Only null left

Chemical Identifiers

CAS 21339-47-9
Molecular Formula C13H24O6
Molecular Weight (g/mol) 276.329
MDL Number MFCD00671525
InChI Key VQUXMFKGQFXVBC-UHFFFAOYSA-N
Synonym 3,3-Diethoxypropane-1,1-dicarboxylic Acid Diethyl Ester
PubChem CID 298328
IUPAC Name diethyl 2-(2,2-diethoxyethyl)propanedioate
SMILES CCOC(CC(C(=O)OCC)C(=O)OCC)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 276.33
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate