accessibility menu, dialog, popup

UserName

CH3OOOH3COOH3COCH3

CAS RN 21339-47-9

IUPAC Name: 1,3-diethyl 2-(2,2-diethoxyethyl)propanedioate
Synonyms: 1,3-diethyl 2-(2,2-diethoxyethyl)propanedioate
Molecular Weight (g/mol): 276.33
Molecular Formula: C13H24O6
InChi Key: VQUXMFKGQFXVBC-UHFFFAOYSA-N
SMILES: CCOC(CC(C(=O)OCC)C(=O)OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results