missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 276.33 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate |
Safety and Handling
| TSCA | No |