missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-Oxopropyl)phosphonate 95.0+%, TCI America™

Catalog No. p-7127614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31745G 5 g
D317425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31745G Supplier TCI America Supplier No. D31745G

Chemical Identifiers

CAS 1067-71-6
Molecular Formula C7H15O4P
Molecular Weight (g/mol) 194.167
MDL Number MFCD00044728
InChI Key RSAFKRSMGOSHRK-UHFFFAOYSA-N
Synonym diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
PubChem CID 14028
IUPAC Name 1-diethoxyphosphorylpropan-2-one
SMILES CCOP(=O)(CC(=O)C)OCC

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 194.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (2-Oxopropyl)phosphonate