missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-oxopropyl)phosphonate, 96%

Catalog No. AC123690250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123690250 25 g Glass bottle
AC123690050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC123690250 Supplier Thermo Scientific Chemicals Supplier No. 123690250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

For C-C Bond Formation (Olefination) | CAS: 1067-71-6 | C7H19O4P | 198.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1067-71-6
Molecular Formula C7H19O4P
Molecular Weight (g/mol) 198.2
MDL Number MFCD00044728
InChI Key RSAFKRSMGOSHRK-UHFFFAOYSA-N
Synonym diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
PubChem CID 14028
IUPAC Name 1-diethoxyphosphorylpropan-2-one
SMILES CCOP(=O)(CC(=O)C)OCC

Specifications

Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 98.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3COCH2P(=O)(OCH2CH3)2
Refractive Index 1.4325 to 1.4355
Quantity 25 g
Fieser 13,255
Specific Gravity 1.11
Formula Weight 198.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl (2-oxopropyl)phosphonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

RUO – Research Use Only