missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-Chlorobenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D52651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52651G 1 g
D52655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52651G Supplier TCI America Supplier No. D52651G

Chemical Identifiers

CAS 29074-98-4
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
MDL Number MFCD22574983
InChI Key GKGLKJMZAVRUQO-UHFFFAOYSA-N
Synonym (2-Chlorobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 12520073
IUPAC Name diethyl [(2-chlorophenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC=CC=C1Cl)OCC

Specifications

Color Colorless
Quantity 1 g
Physical Form Clear Liquid at 20°C
Boiling Point 146°C
Formula Weight 262.67
Percent Purity ≥98.0% (GC)
Chemical Name or Material Diethyl (2-Chlorobenzyl)phosphonate