accessibility menu, dialog, popup

UserName

H3COPOClOCH3

CAS RN 29074-98-4

IUPAC Name: diethyl [(2-chlorophenyl)methyl]phosphonate
Synonyms: diethyl (2-chlorophenyl)methylphosphonate
Molecular Weight (g/mol): 262.67
Molecular Formula: C11H16ClO3P
InChi Key: GKGLKJMZAVRUQO-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=CC=C1Cl)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results