missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Bromoethylphosphonate, TCI America™

Catalog No. B17815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17815G 5 g
B178125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17815G Supplier TCI America Supplier No. B17815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5324-30-1
Molecular Formula C6H14BrO3P
Molecular Weight (g/mol) 245.05
MDL Number MFCD00000239
InChI Key PINITSMLVXAASM-UHFFFAOYSA-N
Synonym diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
PubChem CID 79218
IUPAC Name diethyl (2-bromoethyl)phosphonate
SMILES CCOP(=O)(CCBr)OCC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 245.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2-Bromoethylphosphonate