accessibility menu, dialog, popup

UserName

CH3OPOBrOH3C

CAS RN 5324-30-1

IUPAC Name: diethyl (2-bromoethyl)phosphonate
Synonyms: diethyl 2-bromoethylphosphonate
Molecular Weight (g/mol): 245.05
Molecular Formula: C6H14BrO3P
InChi Key: PINITSMLVXAASM-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCBr)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)