missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-bromoethylphosphonate, 97%

Catalog No. AC114180250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114180250 25 g Glass bottle
AC114180050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114180250 Supplier Thermo Scientific Chemicals Supplier No. 114180250

CAS: 5324-30-1 | C6H14BrO3P | 245.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 5324-30-1
Molecular Formula C6H14BrO3P
Molecular Weight (g/mol) 245.05
MDL Number MFCD00000239
InChI Key PINITSMLVXAASM-UHFFFAOYSA-N
Synonym diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
PubChem CID 79218
SMILES CCOP(=O)(CCBr)OCC

Specifications

Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 75°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4590 to 1.461
Linear Formula (C2H5O)2P(O)CH2CH2Br
Quantity 25 g
Specific Gravity 1.27
IUPAC Name 1-bromo-2-diethoxyphosphorylethane
Formula Weight 245.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl 2-bromoethylphosphonate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-193-3

RUO – Research Use Only