missing translation for 'onlineSavingsMsg'
Learn More

Diethyl [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonate 95.0+%, TCI America™

Catalog No. D42565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42565G 5 g
D42561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42565G Supplier TCI America Supplier No. D42565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116384-53-3
Molecular Formula C12H20N5O4P
Molecular Weight (g/mol) 329.30
MDL Number MFCD07783825
InChI Key SACBMARVYGBCAK-UHFFFAOYSA-N
Synonym 9-[2-(Diethoxyphosphorylmethoxy)ethyl]adenine, [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic Acid Diethyl Ester
PubChem CID 472244
IUPAC Name diethyl {[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}phosphonate
SMILES CCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCC

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 329.30
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonate