accessibility menu, dialog, popup

UserName

CAS RN 116384-53-3

IUPAC Name: diethyl {[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}phosphonate
Synonyms: diethyl [2-(6-aminopurin-9-yl)ethoxy]methylphosphonate
Molecular Weight (g/mol): 329.3
Molecular Formula: C12H20N5O4P
InChi Key: SACBMARVYGBCAK-UHFFFAOYSA-N
SMILES: CCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results