missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 2,5-Dibromoterephthalate 98.0+%, TCI America™
$89.48 - $348.57
Chemical Identifiers
| CAS | 18013-97-3 |
|---|---|
| Molecular Formula | C12H12Br2O4 |
| Molecular Weight (g/mol) | 380.032 |
| MDL Number | MFCD12407163 |
| InChI Key | WXRSDHICEYICMV-UHFFFAOYSA-N |
| Synonym | 2,5-Dibromoterephthalic Acid Diethyl Ester |
| PubChem CID | 22146229 |
| IUPAC Name | diethyl 2,5-dibromobenzene-1,4-dicarboxylate |
| SMILES | CCOC(=O)C1=CC(=C(C=C1Br)C(=O)OCC)Br |
Chemical Identifiers
| 18013-97-3 | |
| 380.032 | |
| WXRSDHICEYICMV-UHFFFAOYSA-N | |
| 22146229 | |
| CCOC(=O)C1=CC(=C(C=C1Br)C(=O)OCC)Br |
| C12H12Br2O4 | |
| MFCD12407163 | |
| 2,5-Dibromoterephthalic Acid Diethyl Ester | |
| diethyl 2,5-dibromobenzene-1,4-dicarboxylate |
Specifications
| 18013-97-3 | |
| White | |
| C12H12Br2O4 | |
| 5 g | |
| WXRSDHICEYICMV-UHFFFAOYSA-N | |
| diethyl 2,5-dibromobenzene-1,4-dicarboxylate | |
| 22146229 | |
| ≥98.0% (GC) | |
| Diethyl 2,5-Dibromoterephthalate |
| 129°C | |
| 335°C | |
| MFCD12407163 | |
| 2,5-Dibromoterephthalic Acid Diethyl Ester | |
| CCOC(=O)C1=CC(=C(C=C1Br)C(=O)OCC)Br | |
| 380.032 | |
| 380.03 | |
| Crystalline Powder |
Safety and Handling
TSCA : No