missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-Pyrrolidinemethylphosphonate 95.0+%, TCI America™

Catalog No. P12585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12585G Supplier TCI America Supplier No. P12585G

Chemical Identifiers

CAS 51868-96-3
Molecular Formula C9H21NO3P
Molecular Weight (g/mol) 222.24
MDL Number MFCD00040516
InChI Key DOMZGECNJWVKAM-UHFFFAOYSA-O
Synonym 1-Pyrrolidinemethylphosphonic Acid Diethyl Ester
PubChem CID 103998
IUPAC Name 1-[(diethoxyphosphoryl)methyl]pyrrolidin-1-ium
SMILES CCOP(=O)(C[NH+]1CCCC1)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 221.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1-Pyrrolidinemethylphosphonate