accessibility menu, dialog, popup

UserName

H3COPONHOCH3

CAS RN 51868-96-3

IUPAC Name: 1-[(diethoxyphosphoryl)methyl]pyrrolidin-1-ium
Synonyms: 1-[(diethoxyphosphoryl)methyl]pyrrolidin-1-ium
Molecular Weight (g/mol): 222.24
Molecular Formula: C9H21NO3P
InChi Key: DOMZGECNJWVKAM-UHFFFAOYSA-O
SMILES: CCOP(=O)(C[NH+]1CCCC1)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results