missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-octylphosphonate, 98%

Catalog No. AAL1858806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1858806 5 g
AAL1858814 25 g
AAL1858822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1858806 Supplier Thermo Scientific Chemicals Supplier No. L1858806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1068-07-1 | C12H27O3P | 250.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1068-07-1
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00015659
InChI Key MVDVRXOWIPONFY-UHFFFAOYSA-N
Synonym diethyl octylphosphonate, diethyl 1-octylphosphonate, phosphonic acid, octyl-, diethyl ester, acmc-1bsbo, octylphosphonic acid diethyl ester, diethyl 1-octylphosphonate, 98+%, octyl-phosphonic acid diethyl ester, phosphonic acid,p-octyl-, diethyl ester, phosphonic acid, p-octyl-, diethyl ester
PubChem CID 70604
IUPAC Name 1-diethoxyphosphoryloctane
SMILES CCCCCCCCP(=O)(OCC)OCC

Specifications

Density 0.953
Boiling Point 88°C to 89°C (0.1 mmHg)
Refractive Index 1.434
Quantity 5 g
Beilstein 1778018
Formula Weight 250.32
Percent Purity ≥98%
Chemical Name or Material Diethyl 1-octylphosphonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 213-941-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only