accessibility menu, dialog, popup

UserName

CAS RN 1068-07-1

IUPAC Name: diethyl octylphosphonate
Synonyms: diethyl octylphosphonate
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H27O3P
InChi Key: MVDVRXOWIPONFY-UHFFFAOYSA-N
SMILES: CCCCCCCCP(=O)(OCC)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results