missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. D37751G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37751G 1 g
D37755G 5 g
D377525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D37751G Supplier TCI America Supplier No. D37751G

Chemical Identifiers

CAS 1149-23-1
Molecular Formula C13H19NO4
Molecular Weight (g/mol) 253.298
MDL Number MFCD00005951
InChI Key LJXTYJXBORAIHX-UHFFFAOYSA-N
Synonym diludine, diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, hantzsch ester, ethidine, etidin, diethone, hantzsch's dihydropyridine, diethyl 2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, diludin, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-, diethyl ester
PubChem CID 70849
IUPAC Name diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1)C(=O)OCC)C)C

Specifications

Melting Point 183°C
Quantity 1 g
Formula Weight 253.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
RTECSNumber US7970100
TSCA No