accessibility menu, dialog, popup

UserName

CAS RN 1149-23-1

IUPAC Name: 3,5-diethyl 2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: etidin
Molecular Weight (g/mol): 253.3
Molecular Formula: C13H19NO4
InChi Key: LJXTYJXBORAIHX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NC(C)=C(C1)C(=O)OCC
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)