accessibility menu, dialog, popup

UserName

H3COOOOH3C

CAS RN 1139-13-5

IUPAC Name: 1,1-diethyl cyclohexane-1,1-dicarboxylate
Synonyms: 1,1-diethyl cyclohexane-1,1-dicarboxylate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: MLHUKQNAQSRVKI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CCCCC1)C(=O)OCC
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results