missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,1-Cyclobutanedicarboxylate 97.0+%, TCI America™
Supplier: TCI America C094225ML
Specifications
| Diethyl 1,1-Cyclobutanedicarboxylate | |
| Yellow | |
| C10H16O4 | |
| 25 mL | |
| JPNJEJSZSMXWSV-UHFFFAOYSA-N | |
| 1,1-diethyl cyclobutane-1,1-dicarboxylate | |
| 77410 | |
| ≥97.0% (GC) |
| 3779-29-1 | |
| 226°C | |
| MFCD00019261 | |
| diethyl 1,1-cyclobutanedicarboxylate, 1,1-cyclobutanedicarboxylic acid, diethyl ester, 1,1-diethyl cyclobutane-1,1-dicarboxylate, cyclobutane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid, 1,1-diethyl ester, 1,1-dicarboethoxycyclobutane, ethyl 1-ethoxycarbonyl cyclobutanecarboxylate, acmc-1cmcx, diethyl1,1-cyclobutanedicarboxylate | |
| CCOC(=O)C1(CCC1)C(=O)OCC | |
| 200.23 | |
| 200.23 | |
| Liquid |
Chemical Identifiers
| 3779-29-1 | |
| 200.23 | |
| JPNJEJSZSMXWSV-UHFFFAOYSA-N | |
| 77410 | |
| CCOC(=O)C1(CCC1)C(=O)OCC |
| C10H16O4 | |
| MFCD00019261 | |
| diethyl 1,1-cyclobutanedicarboxylate, 1,1-cyclobutanedicarboxylic acid, diethyl ester, 1,1-diethyl cyclobutane-1,1-dicarboxylate, cyclobutane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid, 1,1-diethyl ester, 1,1-dicarboethoxycyclobutane, ethyl 1-ethoxycarbonyl cyclobutanecarboxylate, acmc-1cmcx, diethyl1,1-cyclobutanedicarboxylate | |
| 1,1-diethyl cyclobutane-1,1-dicarboxylate |
Safety and Handling
TSCA : Yes