missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1,1-Cyclobutanedicarboxylate 97.0+%, TCI America™

Catalog No. p-7124697
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09425ML 5 mL
C094225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09425ML Supplier TCI America Supplier No. C09425ML
Only null left

Chemical Identifiers

CAS 3779-29-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00019261
InChI Key JPNJEJSZSMXWSV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclobutanedicarboxylate, 1,1-cyclobutanedicarboxylic acid, diethyl ester, 1,1-diethyl cyclobutane-1,1-dicarboxylate, cyclobutane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid diethyl ester, 1,1-cyclobutanedicarboxylic acid, 1,1-diethyl ester, 1,1-dicarboethoxycyclobutane, ethyl 1-ethoxycarbonyl cyclobutanecarboxylate, acmc-1cmcx, diethyl1,1-cyclobutanedicarboxylate
PubChem CID 77410
IUPAC Name 1,1-diethyl cyclobutane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CCC1)C(=O)OCC

Specifications

Color Yellow
Boiling Point 226°C
Quantity 5 mL
Formula Weight 200.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 1,1-Cyclobutanedicarboxylate