missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethoxymethyl Acetate 97.0+%, TCI America™
$136.08 - $456.70
Chemical Identifiers
| CAS | 14036-06-7 |
|---|---|
| Molecular Formula | C7H14O4 |
| Molecular Weight (g/mol) | 162.185 |
| MDL Number | MFCD00009229 |
| InChI Key | IRUNKQSGDBYUDC-UHFFFAOYSA-N |
| Synonym | methanol, diethoxy-, acetate, diethoxymethylacetate, acetic acid diethoxymethyl ester, methanol, 1,1-diethoxy-, 1-acetate, methyl2,2-diethoxyacetate, pubchem13756, acmc-209ckk, diethoxymethyl acetate, qspl 099, diethoxy-acetic acid methyl ester |
| PubChem CID | 84166 |
| IUPAC Name | diethoxymethyl acetate |
| SMILES | CCOC(OCC)OC(=O)C |
Chemical Identifiers
| 14036-06-7 | |
| 162.185 | |
| IRUNKQSGDBYUDC-UHFFFAOYSA-N | |
| 84166 | |
| CCOC(OCC)OC(=O)C |
| C7H14O4 | |
| MFCD00009229 | |
| methanol, diethoxy-, acetate, diethoxymethylacetate, acetic acid diethoxymethyl ester, methanol, 1,1-diethoxy-, 1-acetate, methyl2,2-diethoxyacetate, pubchem13756, acmc-209ckk, diethoxymethyl acetate, qspl 099, diethoxy-acetic acid methyl ester | |
| diethoxymethyl acetate |
Specifications
| 14036-06-7 | |
| C7H14O4 | |
| 5 g | |
| methanol, diethoxy-, acetate, diethoxymethylacetate, acetic acid diethoxymethyl ester, methanol, 1,1-diethoxy-, 1-acetate, methyl2,2-diethoxyacetate, pubchem13756, acmc-209ckk, diethoxymethyl acetate, qspl 099, diethoxy-acetic acid methyl ester | |
| CCOC(OCC)OC(=O)C | |
| 162.185 | |
| 162.19 | |
| Liquid |
| Yellow | |
| MFCD00009229 | |
| 3272 | |
| IRUNKQSGDBYUDC-UHFFFAOYSA-N | |
| diethoxymethyl acetate | |
| 84166 | |
| ≥97.0% (GC) | |
| Diethoxymethyl Acetate |
Safety and Handling
TSCA : Yes