accessibility menu, dialog, popup

UserName

CAS RN 14036-06-7

IUPAC Name: diethoxymethyl acetate
Synonyms: diethoxymethyl acetate
Molecular Weight (g/mol): 162.19
Molecular Formula: C7H14O4
InChi Key: IRUNKQSGDBYUDC-UHFFFAOYSA-N
SMILES: CCOC(OCC)OC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results