missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethyl Acetate 97.0+%, TCI America™

Catalog No. A16085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16085G 5 g
A160825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16085G Supplier TCI America Supplier No. A16085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14036-06-7
Molecular Formula C7H14O4
Molecular Weight (g/mol) 162.185
MDL Number MFCD00009229
InChI Key IRUNKQSGDBYUDC-UHFFFAOYSA-N
Synonym methanol, diethoxy-, acetate, diethoxymethylacetate, acetic acid diethoxymethyl ester, methanol, 1,1-diethoxy-, 1-acetate, methyl2,2-diethoxyacetate, pubchem13756, acmc-209ckk, diethoxymethyl acetate, qspl 099, diethoxy-acetic acid methyl ester
PubChem CID 84166
IUPAC Name diethoxymethyl acetate
SMILES CCOC(OCC)OC(=O)C

Specifications

Color Yellow
Quantity 5 g
UN Number 3272
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxymethyl Acetate