missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydiphenylsilane 98.0+%, TCI America™

Catalog No. p-7127601
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D306725G 25 g
D3067100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D306725G Supplier TCI America Supplier No. D306725G
Only null left

Chemical Identifiers

CAS 2553-19-7
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.42
MDL Number MFCD00015126
InChI Key ZZNQQQWFKKTOSD-UHFFFAOYSA-N
Synonym diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
PubChem CID 75705
IUPAC Name diethoxydiphenylsilane
SMILES CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 218°C
Quantity 25 g
Formula Weight 272.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxydiphenylsilane
EINECSNumber (3)-2635
TSCA Yes