missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, Reagent, ACS, 98.5%, Spectrum™ Chemical

Catalog No. p-7157297
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602183 100 mL Amber Glass Bottle
18602184 500 mL Amber Glass Bottle
18602185 6 x 500 mL Amber Glass Bottle
18602186 4 L Amber Glass Bottle
18602187 4 x 4 L Amber Glass Bottle
18602188 20 L Steel Can
18602189 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18602183 Supplier Spectrum Chemical Mfg Cor Supplier No. D1033100ML
Only null left

D1033, 111-42-2, C4H11NO2

Spectrum™ Chemical Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 98.5%
Grade ACS