missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, 99%

Catalog No. p-3736221
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113920010 1 kg Glass Bottle
AC113920025 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113920010 Supplier Thermo Scientific Chemicals Supplier No. 113920010
Only null left

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Density 1.0955g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Refractive Index 1.4760 to 1.4790 (20°C, 589nm)
Linear Formula (HOCH2CH2)2NH
Quantity 1 kg
Beilstein 04, 283
Merck Index 15, 3118
Vapor Pressure 2hPa at 10°C
Formula Weight 105.14
Percent Purity ≥98.5%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Diethanolamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Risk of serious damage to eyes. May cause irritation of respiratory tract. Corrosive to metals.
NFPA&C

RUO – Research Use Only