missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine, 99%

Catalog No. AAA1338930
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1338930 250 g
AAA1338936 500 g
AAA133890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1338930 Supplier Thermo Scientific Chemicals Supplier No. A1338930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

Solubility
Miscible with water.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, strong acids, copper, zinc, carbon dioxide and iron.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Melting Point 27°C to 30°C
Density 1.097
pH 11
Boiling Point 268°C to 270°C
Flash Point 176°C (349°F)
Odor Ammonia-like
Assay Percent Range 99%
Refractive Index 1.477
Quantity 250 g
Beilstein 605315
Sensitivity Hygroscopic
Merck Index 14,3107
Solubility Information Miscible with water.
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material Diethanolamine
Show More Show Less
Hazard Category H302-H315-H318-H335-H351-H373
Hazard Statement GHS H Statement
H318-H373-H302-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P201-P202-P234-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-868-0
RTECSNumber KL2975000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only