missing translation for 'onlineSavingsMsg'
Learn More

Didecyl Phosphate 95.0+%, TCI America™

Catalog No. P1011100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1011100MG 100 mg
P10111G 1 g
P10115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1011100MG Supplier TCI America Supplier No. P1011100MG

Chemical Identifiers

CAS 7795-87-1
Molecular Formula C20H43O4P
Molecular Weight (g/mol) 378.53
MDL Number MFCD00015062
InChI Key QHAUASBJFFBWMY-UHFFFAOYSA-N
Synonym Phosphoric Acid Didecyl Ester
PubChem CID 82260
IUPAC Name bis(decyloxy)phosphinic acid
SMILES CCCCCCCCCCOP(O)(=O)OCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Quantity 100 mg
Formula Weight 378.53
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didecyl Phosphate
EINECSNumber (2)-1986
TSCA Yes