accessibility menu, dialog, popup

UserName

H3COPOHOOCH3

CAS RN 7795-87-1

IUPAC Name: bis(decyloxy)phosphinic acid
Synonyms: bis(decyloxy)phosphinic acid
Molecular Weight (g/mol): 378.53
Molecular Formula: C20H43O4P
InChi Key: QHAUASBJFFBWMY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOP(O)(=O)OCCCCCCCCCC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results