missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopropylmethanol, 97%

Catalog No. AAL0181603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0181603 1 g
AAL0181606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0181603 Supplier Thermo Scientific Chemicals Supplier No. L0181603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14300-33-5 | C7H12O | 112.17 g/mol

Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopropylmethanol is used as protecting group reagent for carboxylic acids; the formed esters are very mildly and selectively cleaved.

Solubility
Sparingly soluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14300-33-5
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00019249
InChI Key PIXLZMHERIHLJL-UHFFFAOYSA-N
Synonym dicyclopropyl carbinol, dicyclopropylmethyl alcohol, dicyclopropylmethan-1-ol, cyclopropanemethanol, a-cyclopropyl, cyclopropanemethanol, .alpha.-cyclopropyl, dicyclopropyl-methanol, acmc-20ala1, alpha-cyclopropylcyclopropanemethanol
PubChem CID 84336
IUPAC Name dicyclopropylmethanol
SMILES OC(C1CC1)C1CC1

Specifications

Boiling Point 99°C to 100°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.465
Quantity 1 g
Beilstein 2203215
Solubility Information Sparingly soluble in water.
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material Dicyclopropylmethanol
EINECSNumber 238-236-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only