CAS RN 14300-33-5
IUPAC Name:
dicyclopropylmethanol
Synonyms:
dicyclopropyl carbinol
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
PIXLZMHERIHLJL-UHFFFAOYSA-N
SMILES:
OC(C1CC1)C1CC1
1
–
1
of
1
results
1
Promotions Available
Dicyclopropylmethanol, 97%
Dicyclopropylmethanol, 97%
CAS: 14300-33-5 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.17 MDL Number: MFCD00019249 InChI Key: PIXLZMHERIHLJL-UHFFFAOYSA-N Synonym: dicyclopropyl carbinol,dicyclopropylmethyl alcohol,dicyclopropylmethan-1-ol,cyclopropanemethanol, a-cyclopropyl,cyclopropanemethanol, .alpha.-cyclopropyl,dicyclopropyl-methanol,acmc-20ala1,alpha-cyclopropylcyclopropanemethanol PubChem CID: 84336 IUPAC Name: dicyclopropylmethanol SMILES: OC(C1CC1)C1CC1