missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopropyl Ketone 95.0+%, TCI America™

Catalog No. D175825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D175825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D175825ML Supplier TCI America Supplier No. D175825ML

Chemical Identifiers

CAS 1121-37-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00001276
InChI Key BIPUHAHGLJKIPK-UHFFFAOYSA-N
Synonym dicyclopropyl ketone, cyclopropyl ketone, dicyclopropylketone, methanone, dicyclopropyl, dicyclopropyl-methanone, dicyclopropylmethyloxime, dicyclopropyl methyl ketone, pubchem13596, methanone,dicyclopropyl, acmc-1busz
PubChem CID 70721
IUPAC Name dicyclopropylmethanone
SMILES C1CC1C(=O)C2CC2

Specifications

Boiling Point 67°C
Quantity 25 mL
UN Number 1224
Formula Weight 110.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclopropyl Ketone