accessibility menu, dialog, popup

UserName

O

CAS RN 1121-37-5

IUPAC Name: dicyclopropylmethanone
Synonyms: methanone, dicyclopropyl-
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: BIPUHAHGLJKIPK-UHFFFAOYSA-N
SMILES: O=C(C1CC1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results