missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclopentanyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™
$65.24 - $417.56
Chemical Identifiers
| CAS | 7398-56-3 |
|---|---|
| Molecular Formula | C13H18O2 |
| Molecular Weight (g/mol) | 206.29 |
| MDL Number | MFCD08276368 |
| InChI Key | BDYXPMNDZDGGTI-UHFFFAOYNA-N |
| Synonym | Acrylic Acid Dicyclopentanyl Ester, Acrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Acrylate |
| PubChem CID | 13001872 |
| IUPAC Name | tricyclo[5.2.1.0²,â¶]decan-8-yl prop-2-enoate |
| SMILES | C=CC(=O)OC1CC2CC1C1CCCC21 |
Chemical Identifiers
| 7398-56-3 | |
| 206.29 | |
| BDYXPMNDZDGGTI-UHFFFAOYNA-N | |
| 13001872 | |
| C=CC(=O)OC1CC2CC1C1CCCC21 |
| C13H18O2 | |
| MFCD08276368 | |
| Acrylic Acid Dicyclopentanyl Ester, Acrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Acrylate | |
| tricyclo[5.2.1.0²,â¶]decan-8-yl prop-2-enoate |
Specifications
| 7398-56-3 | |
| C13H18O2 | |
| 25 g | |
| BDYXPMNDZDGGTI-UHFFFAOYNA-N | |
| tricyclo[5.2.1.0²,â¶]decan-8-yl prop-2-enoate | |
| 13001872 | |
| ≥95.0% (GC) | |
| Dicyclopentanyl Acrylate (stabilized with MEHQ) |
| Colorless | |
| MFCD08276368 | |
| Acrylic Acid Dicyclopentanyl Ester, Acrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Acrylate | |
| C=CC(=O)OC1CC2CC1C1CCCC21 | |
| 206.29 | |
| 206.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-1583
TSCA : No
Recommended Storage : Freezer