missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene (stabilized with BHT) 97.0+%, TCI America™

Catalog No. p-7126216
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D044325ML 25 mL
D0443500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D044325ML Supplier TCI America Supplier No. D044325ML
Only null left

Chemical Identifiers

CAS 77-73-6
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00078246
InChI Key HECLRDQVFMWTQS-UHFFFAOYSA-N
Synonym dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
PubChem CID 6492
ChEBI CHEBI:34695
SMILES C1C=CC2C1C3CC2C=C3

Specifications

Color White
Boiling Point 167°C
Quantity 25 mL
UN Number 2048
Formula Weight 132.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dicyclopentadiene (stabilized with BHT) [precursor to Cyclopentadiene]
EINECSNumber (4)-0634
RTECSNumber PC1050000
TSCA Yes