accessibility menu, dialog, popup

UserName

CAS RN 77-73-6

IUPAC Name: tricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms: dicyclopentadiene 1,3-cyclopentadiene, dimer dimer 1,3-cyclopentadiene dicyklopentadien dimer cyklopentadienu bicyclopentadiene biscyclopentadiene cyclopentadiene dimer
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: HECLRDQVFMWTQS-UHFFFAOYNA-N
SMILES: C1C=CC2C3CC(C=C3)C12
Boiling Point 
  • (2)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (8)
  • (2)
  • (4)
  • (4)
  • (4)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)