missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentadiene diepoxide, 98%

Catalog No. p-7043827
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0294006 5 g
AAL0294014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0294006 Supplier Thermo Scientific Chemicals Supplier No. L0294006
Only null left

CAS: 81-21-0 | C10H12O2 | 164.204 g/mol

Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene diepoxide acts as an intermediate for epoxy resins, plasticizers and protective coatings. It finds application as a rubber additive and adhesives, which is used at high temperature. Further, it is used in the preparation of endo-3-hydroxy-exo-9-oxa-exo-tetracyclo[5.3.1.02,6.08,10]undecane using lithium aluminum hydride as a reagent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 81-21-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00077209
InChI Key BQQUFAMSJAKLNB-UHFFFAOYSA-N
Synonym dicyclopentadiene dioxide, dicyclopentadiene diepoxide, unox epoxide 207, epoxide 207, bicyclopentadiene dioxide, unox 207x, unox 207, dicyclopentadiene dioxide van, 1,2:5,6-diepoxyhexahydro-4,7-methanoindan, 4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
PubChem CID 6673
SMILES C1C2C3CC4C(C3C1C5C2O5)O4

Specifications

Melting Point 185°C to 189°C
Density 1.331
Boiling Point 326°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 112743
Solubility Information Soluble in water.
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material Dicyclopentadiene diepoxide
Show More Show Less
Hazard Category H301-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-334-1
RTECSNumber PB9625200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only