missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylphenylphosphine 95.0+%, TCI America™

Catalog No. p-7127283
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24111G 1 g
D24115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24111G Supplier TCI America Supplier No. D24111G
Only null left

Chemical Identifiers

CAS 6476-37-5
Molecular Formula C18H27P
Molecular Weight (g/mol) 274.39
MDL Number MFCD00003854
InChI Key VPLLTGLLUHLIHA-UHFFFAOYSA-N
Synonym dicyclohexylphenylphosphine, dicyclohexyl phenyl phosphine, phosphine, dicyclohexylphenyl, phenyldicyclohexylphosphine, phenylphosphinodicyclohexane, dicyclohexyl phenyl phosphane, pphcy2, acmc-1bey7, dicyclohexyl-phenyl-phosphane
PubChem CID 80970
IUPAC Name dicyclohexyl(phenyl)phosphane
SMILES C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 58°C
Color White
Quantity 1 g
Formula Weight 274.39
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylphenylphosphine
RTECSNumber SY9125000
TSCA No