CAS RN 6476-37-5
CAS RN 6476-37-5
IUPAC Name:
dicyclohexyl(phenyl)phosphane
Synonyms:
dicyclohexylphenylphosphine
Molecular Weight (g/mol):
274.39
Molecular Formula:
C18H27P
InChi Key:
VPLLTGLLUHLIHA-UHFFFAOYSA-N
SMILES:
C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
Dicyclohexylphenylphosphine 95.0+%, TCI America™
CAS: 6476-37-5 Molecular Formula: C18H27P Molecular Weight (g/mol): 274.39 MDL Number: MFCD00003854 InChI Key: VPLLTGLLUHLIHA-UHFFFAOYSA-N Synonym: dicyclohexylphenylphosphine,dicyclohexyl phenyl phosphine,phosphine, dicyclohexylphenyl,phenyldicyclohexylphosphine,phenylphosphinodicyclohexane,dicyclohexyl phenyl phosphane,pphcy2,acmc-1bey7,dicyclohexyl-phenyl-phosphane PubChem CID: 80970 IUPAC Name: dicyclohexyl(phenyl)phosphane SMILES: C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1