missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylmethane 4,4'-Diisocyanate (mixture of isomers) 90.0+%, TCI America™

Catalog No. D238025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238025G 25 g
D2380100G 100 g
D2380500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D238025G Supplier TCI America Supplier No. D238025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5124-30-1
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD00015405
InChI Key KORSJDCBLAPZEQ-UHFFFAOYSA-N
Synonym hydrogenated mdi, hylene w, bis 4-isocyanatocyclohexyl methane, 4,4'-diisocyanatodicyclohexylmethane, methylene bis 4-cyclohexylisocyanate, dicyclohexylmethane 4,4'-diisocyanate, dicyclohexylmethane-4,4'-diisocyanate, methylene bis-4-cyclohexylisocyanate, nacconate h 12, cyclohexane, 1,1'-methylenebis 4-isocyanato
PubChem CID 21202
ChEBI CHEBI:53216
IUPAC Name 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
SMILES O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
UN Number 2206
Formula Weight 262.35
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclohexylmethane 4,4′-Diisocyanate (mixture of isomers)
EINECSNumber (4)-0119
RTECSNumber NQ9250000
TSCA Yes