missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl Phthalate 99.0+%, TCI America™

Catalog No. P029325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P029325G 25 g
P0293500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P029325G Supplier TCI America Supplier No. P029325G

Chemical Identifiers

CAS 84-61-7
Molecular Formula C20H26O4
Molecular Weight (g/mol) 330.424
MDL Number MFCD00003849
InChI Key VOWAEIGWURALJQ-UHFFFAOYSA-N
Synonym dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190
PubChem CID 6777
ChEBI CHEBI:34693
IUPAC Name dicyclohexyl benzene-1,2-dicarboxylate
SMILES C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3

Specifications

Melting Point 65°C
Color White
Boiling Point 235°C
Quantity 25 g
Formula Weight 330.42
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexyl Phthalate
EINECSNumber (3)-1311
RTECSNumber TI0889000
TSCA Yes