Learn More
Dicyclohexyl phthalate, 99%
CAS: 84-61-7 | C20H26O4 | 330.42 g/mol
$48.24 - $48.24
Chemical Identifiers
| CAS | 84-61-7 |
|---|---|
| Molecular Formula | C20H26O4 |
| Molecular Weight (g/mol) | 330.42 |
| MDL Number | MFCD00003849 |
| InChI Key | VOWAEIGWURALJQ-UHFFFAOYSA-N |
| Synonym | dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 |
| PubChem CID | 6777 |
| ChEBI | CHEBI:34693 |
| IUPAC Name | dicyclohexyl benzene-1,2-dicarboxylate |
| SMILES | C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC375050050
|
Thermo Scientific Chemicals
375050050 |
5 g | Glass bottle |
Each for $48.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 84-61-7 | |
| 330.42 | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| 6777 | |
| dicyclohexyl benzene-1,2-dicarboxylate |
| C20H26O4 | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| CHEBI:34693 | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
Specifications
| 84-61-7 | |
| White | |
| 200.0°C to 235.0°C (4.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| dicyclohexyl benzene-1,2-dicarboxylate | |
| 6777 | |
| 330.42 | |
| Crystalline Powder |
| 63.0°C | |
| 207g/mL | |
| 207°C | |
| 98.5% min. (GC) | |
| C20H26O4 | |
| 5 g | |
| Solubility in water: insoluble | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 | |
| 330.42 | |
| CHEBI:34693 | |
| 99% | |
| Dicyclohexyl phthalate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May damage the unborn child.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention
GHS Signal Word: Danger
EINECSNumber : 201-545-9
RUO – Research Use Only