missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl Ketone 97.0+%, TCI America™
$31.68 - $260.97
Chemical Identifiers
| CAS | 119-60-8 |
|---|---|
| Molecular Formula | C13H22O |
| Molecular Weight (g/mol) | 194.318 |
| MDL Number | MFCD00040417 |
| InChI Key | TWXWPPKDQOWNSX-UHFFFAOYSA-N |
| Synonym | Dodecahydrobenzophenone |
| PubChem CID | 8403 |
| IUPAC Name | dicyclohexylmethanone |
| SMILES | C1CCC(CC1)C(=O)C2CCCCC2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D225825ML
|
TCI America
D225825ML |
25 mL |
Each for $31.68
|
|
|||||
|
D2258500ML
|
TCI America
D2258500ML |
500 mL |
Each for $260.97
|
|
|||||
Chemical Identifiers
| 119-60-8 | |
| 194.318 | |
| TWXWPPKDQOWNSX-UHFFFAOYSA-N | |
| 8403 | |
| C1CCC(CC1)C(=O)C2CCCCC2 |
| C13H22O | |
| MFCD00040417 | |
| Dodecahydrobenzophenone | |
| dicyclohexylmethanone |
Specifications
| 119-60-8 | |
| 135°C | |
| MFCD00040417 | |
| Dodecahydrobenzophenone | |
| C1CCC(CC1)C(=O)C2CCCCC2 | |
| 194.318 | |
| 194.32 | |
| Liquid |
| Yellow | |
| C13H22O | |
| 25 mL | |
| TWXWPPKDQOWNSX-UHFFFAOYSA-N | |
| dicyclohexylmethanone | |
| 8403 | |
| ≥97.0% (GC) | |
| Dicyclohexyl Ketone |
Safety and Handling
RTECSNumber : OB1804500
TSCA : Yes