missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl Disulfide 90.0+%, TCI America™

Catalog No. D161725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D161725ML 25 mL
D1617500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D161725ML Supplier TCI America Supplier No. D161725ML

Chemical Identifiers

CAS 2550-40-5
Molecular Formula C12H22S2
Molecular Weight (g/mol) 230.428
MDL Number MFCD00013759
InChI Key ODHAQPXNQDBHSH-UHFFFAOYSA-N
Synonym dicyclohexyl disulfide, cyclohexyl disulfide, disulfide, dicyclohexyl, 1,2-dicyclohexyldisulfane, dicyclohexyl disulphide, bis cyclohexyl disulfide, cyclohexyldisulfanyl cyclohexane, cyclohexyldisulfide, unii-6g1a0k504q
PubChem CID 17356
IUPAC Name (cyclohexyldisulfanyl)cyclohexane
SMILES C1CCC(CC1)SSC2CCCCC2

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
Formula Weight 230.43
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclohexyl Disulfide