missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexano-18-crown 6-Ether 98.0+%, TCI America™

Catalog No. p-7126865
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16681G 1 g
D16685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16681G Supplier TCI America Supplier No. D16681G
Only null left

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.502
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

Melting Point 51°C
Color White
Boiling Point 344°C
Quantity 1 g
UN Number 2811
Formula Weight 372.50
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Dicyclohexano-18-crown 6-Ether
RTECSNumber HP5385000
TSCA Yes