accessibility menu, dialog, popup

UserName

CAS RN 16069-36-6

IUPAC Name: 2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosane
Synonyms: dicyclohexyl-18-crown-6
Molecular Weight (g/mol): 372.5
Molecular Formula: C20H36O6
InChi Key: BBGKDYHZQOSNMU-UHFFFAOYNA-N
SMILES: C1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)